siRNA Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012707.1 | - | 2 | 309 | ||
| AK140734.1 | - | 3 | 229 | |||
| AK220466.1 | - | 3 | 238 | |||
| BC021396.1 | - | 2 | 268 | |||
| BC070412.1 | - | 4 | 403 | 5705112 | ||
| U96963.1 | - | 2 | 241 | |||

Diap1
Genomic Map