siRNA Details
| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007865.3 | NP_031891.2 | 2 | 681 | ||
| GenBank mRNA | BC057400.1 | - | 2 | 194 | 6402691 | |
| BC065063.1 | - | 2 | 445 | 6416191 | ||
| BQ768900.1 | - | 2 | 681 | |||
| X80903.1 | - | 2 | 111 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_032063, U78889.

Dll1
Genomic Map