siRNA Details
| Gene Symbol: Dnajc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190817.1 | NP_001177746.1 | 1 | 509 | ||
| NM_007869.3 | NP_031895.1 | 1 | 509 | |||
| GenBank mRNA | AK012505.1 | - | 1 | 503 | ||
| AK052022.1 | - | 1 | 179 | |||
| AK133582.1 | - | 1 | 493 | |||
| AK144228.1 | - | 1 | 465 | |||
| BC020071.1 | - | 394 | ||||
| BC080300.1 | - | 1 | 490 | |||
| L16953.1 | - | 1 | 186 | |||

Dnajc1
Genomic Map