siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162465.1 | NP_001155937.1 | 5 | 639 | ||
| NM_007886.2 | NP_031912.2 | 5 | 574 | |||
| GenBank mRNA | AJ003007.1 | - | 5 | 507 | ||
| AK043436.1 | - | 5 | 597 | |||
| AK083752.1 | - | 5 | 595 | |||
| AK085675.1 | - | 5 | 564 | |||
| AK157158.1 | - | 5 | 596 | |||
| BC030924.1 | - | 5 | 574 | 4910166 | ||
| Y15742.1 | - | 5 | 471 | |||

Dtnb
Genomic Map