siRNA Details
| Gene Symbol: Cyp2c29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007815.3 | NP_031841.3 | 3, 4 | 511 | ||
| GenBank mRNA | AI182545.1 | - | 3, 4 | 522 | ||
| AK139760.1 | - | 551 | ||||
| AK149558.1 | - | 3, 4 | 330 | |||
| BC019908.1 | - | 3, 4 | 493 | 5123631 | ||
| D17674.1 | - | 3, 4 | 485 | |||

Cyp2c29
Genomic Map