siRNA Details
| Gene Symbol: Dnajc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190817.1 | NP_001177746.1 | 3 | 677 | ||
| NM_007869.3 | NP_031895.1 | 3 | 677 | |||
| GenBank mRNA | AK133582.1 | - | 3 | 661 | ||
| AK144228.1 | - | 3 | 633 | |||
| BC080300.1 | - | 3 | 658 | |||
| L16953.1 | - | 3 | 354 | |||

Dnajc1
Genomic Map