siRNA Details
| Gene Symbol: Arid3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288625.1 | NP_001275554.1 | 2 | 678 | ||
| NM_001288626.1 | NP_001275555.1 | 2 | 832 | |||
| NM_007880.4 | NP_031906.1 | 2 | 832 | |||
| GenBank mRNA | AK141283.1 | - | 2 | 679 | ||
| AK154956.1 | - | 2 | 804 | |||
| BC050925.1 | - | 2 | 736 | 6831312 | ||
| U60335.1 | - | 1 | 620 | |||

Arid3a
Genomic Map