siRNA Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007884.2 | NP_031910.1 | 4 | 317 | ||
| GenBank mRNA | AK005684.1 | - | 4 | 318 | ||
| AK015223.1 | - | 4 | 317 | |||
| AK137179.1 | - | 3 | 242 | |||
| AK157900.1 | - | 568 | ||||
| AK157901.1 | - | 3 | 579 | |||
| BC012695.1 | - | 3 | 289 | 4037028 | ||
| D78274.1 | - | 3 | 232 | |||

Epyc
Genomic Map