siRNA Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 8 | 769 | ||
| GenBank mRNA | AF035527.1 | - | 7 | 767 | ||
| AK028651.1 | - | 8 | 740 | |||
| BC005520.1 | - | 9 | 686 | 3494644 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehf| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 8 | 769 | ||
| GenBank mRNA | AF035527.1 | - | 7 | 767 | ||
| AK028651.1 | - | 8 | 740 | |||
| BC005520.1 | - | 9 | 686 | 3494644 | ||