siRNA Details
| Gene Symbol: Ei24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199494.1 | NP_001186423.1 | 7 | 733 | ||
| NM_007915.5 | NP_031941.1 | 7 | 617 | |||
| GenBank mRNA | AK145018.1 | - | 7 | 694 | ||
| AK147776.1 | - | 7 | 646 | |||
| AK159927.1 | - | 7 | 675 | |||
| AK172233.1 | - | 7 | 697 | |||
| AK212828.1 | - | 2 | 178 | |||
| BU148129.1 | - | 7 | 686 | |||
| U41751.2 | - | 7 | 621 | |||

Ei24
Genomic Map