siRNA Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007916.2 | NP_031942.2 | 7, 8 | 703 | ||
| GenBank mRNA | AK049467.1 | - | 7, 8 | 674 | ||
| AK146548.1 | - | 7, 8 | 664 | |||
| BC011270.1 | - | 7, 8 | 675 | 3674608 | ||
| L25125.1 | - | 7, 8 | 700 | |||

Ddx19a
Genomic Map