siRNA Details
| Gene Symbol: Ddx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110826.1 | NP_001104296.1 | 8 | 1154 | ||
| NM_007841.4 | NP_031867.1 | 8 | 1097 | |||
| NM_181324.3 | NP_851841.2 | 8 | 1122 | |||
| GenBank mRNA | AF038995.1 | - | 7 | 716 | ||
| AK036648.1 | - | 8 | 1150 | |||
| AK054144.1 | - | 8 | 1116 | |||
| AK148483.1 | - | 8 | 1119 | |||
| BC021452.1 | - | 7 | 1014 | 5043599 | ||

Ddx6
Genomic Map