siRNA Details
| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291809.1 | NP_001278738.1 | 9 | 1372 | ||
| NM_007882.3 | NP_031908.3 | 9 | 1372 | |||
| GenBank mRNA | BC144845.1 | - | 9 | 1357 | ||
| CD773111.1 | - | 1 | 25 | |||
| Y11169.1 | - | 9 | 1372 | |||

Dsc3
Genomic Map