siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162465.1 | NP_001155937.1 | 18 | 2334 | ||
| NM_007886.2 | NP_031912.2 | 19 | 2305 | |||
| GenBank mRNA | AJ003007.1 | - | 21 | 2300 | ||
| AK019068.1 | - | 4 | 387 | |||
| AK191798.1 | - | 1 | 263 | |||
| BC030924.1 | - | 19 | 2305 | 4910166 | ||
| Y15742.1 | - | 18 | 2165 | |||

Dtnb
Genomic Map