siRNA Details
| Gene Symbol: Eif4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007917.3 | NP_031943.3 | 8 | 1787 | ||
| GenBank mRNA | AK082867.1 | - | 7 | 1668 | ||
| AK146757.1 | - | 7 | 1667 | |||
| BC085087.1 | - | 7 | 1659 | 6438419 | ||
| M61731.1 | - | 7 | 1673 | |||

Eif4e
Genomic Map