siRNA Details
| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007936.3 | NP_031962.2 | 3 | 230 | ||
| GenBank mRNA | AK028509.1 | - | 3 | 222 | ||
| AK132203.1 | - | 3 | 229 | |||
| AK147698.1 | - | 3 | 226 | |||
| BC004782.1 | - | 3 | 145 | 3588285 | ||
| BC052164.1 | - | 3 | 202 | 6512978 | ||
| S57168.1 | - | 3 | 230 | |||
| X65138.1 | - | 3 | 230 | |||

Epha4
Genomic Map