siRNA Details
| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007969.4 | NP_031995.3 | 3 | 465 | ||
| GenBank mRNA | AK145032.1 | - | 3 | 361 | ||
| BC010986.1 | - | 3 | 465 | 4038377 | ||
| BE310436.1 | - | 3 | 326 | |||
| EH099462.1 | - | 3 | 492 | |||
| EH105860.1 | - | 3 | 492 | |||
| EH109450.1 | - | 3 | 371 | |||
| X93037.1 | - | 3 | 363 | |||

Wfdc18
Genomic Map