siRNA Details
| Gene Symbol: Fabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007980.3 | NP_032006.1 | 2 | 435 | ||
| GenBank mRNA | AA245078.1 | - | 2 | 435 | ||
| AK007995.1 | - | 2 | 133 | |||
| AY523818.1 | - | 2 | 121 | |||
| AY523819.1 | - | 2 | 121 | |||
| BC013457.1 | - | 2 | 115 | 4218050 | ||
| BY707992.1 | - | 2 | 131 | |||
| M65034.1 | - | 2 | 129 | |||

Fabp2
Genomic Map