siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 2 | 420 | ||
| NM_007985.3 | NP_032011.2 | 2 | 420 | |||
| GenBank mRNA | AK035488.1 | - | 2 | 174 | ||
| AK042226.1 | - | 2 | 388 | |||
| AK080697.1 | - | 400 | ||||
| AK147025.1 | - | 2 | 400 | |||
| AK163161.1 | - | 2 | 421 | |||
| AK196042.1 | - | 2 | 150 | |||
| L08266.1 | - | 2 | 401 | |||

Fancc
Genomic Map