siRNA Details
| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146708.1 | NP_001140180.1 | 2 | 131 | ||
| NM_007987.2 | NP_032013.2 | 2 | 131 | |||
| GenBank mRNA | AK002590.1 | - | 2 | 82 | ||
| AK086933.1 | - | 2 | 108 | |||
| BC061160.1 | - | 2 | 104 | 30302649 | ||
| BY348234.1 | - | 2 | 132 | |||
| DQ846748.1 | - | 2 | 75 | |||
| DQ846749.1 | - | 2 | 75 | |||
| L25336.1 | - | 1 | 54 | |||
| M83649.1 | - | 2 | 124 | |||

Fas
Genomic Map