siRNA Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007995.3 | NP_032021.1 | 1, 2 | 222 | ||
| GenBank mRNA | AB007813.1 | - | 1, 2 | 114 | ||
| AB222271.1 | - | 1, 2 | 24 | |||
| AK133539.1 | - | 1, 2 | 199 | |||
| AK138752.1 | - | 1, 2 | 169 | |||
| AK159651.1 | - | 1, 2 | 192 | |||
| BC019180.1 | - | 1, 2 | 170 | 5008934 | ||
| CT010383.1 | - | 1, 2 | 84 | |||

Fcna
Genomic Map