siRNA Details
| Gene Symbol: Elk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007922.5 | NP_031948.4 | 3 | 635 | ||
| GenBank mRNA | AK132354.1 | - | 3 | 628 | ||
| BC054474.1 | - | 3 | 619 | 30045506 | ||
| CV561936.1 | - | 2 | 581 | |||
| X87257.1 | - | 3 | 905 | |||
| Z36939.1 | - | 2 | 420 | |||

Elk1
Genomic Map