siRNA Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271587.1 | NP_001258516.1 | 4 | 475 | ||
| NM_001271588.1 | NP_001258517.1 | 4 | 406 | |||
| NM_001271595.1 | NP_001258524.1 | 4 | 480 | |||
| NM_007945.3 | NP_031971.2 | 5 | 481 | |||
| GenBank mRNA | AK147877.1 | - | 5 | 409 | ||
| AK148424.1 | - | 5 | 397 | |||
| AK149683.1 | - | 4 | 269 | |||
| AK155022.1 | - | 4 | 241 | |||
| AK166156.1 | - | 5 | 316 | |||
| BC016890.1 | - | 4 | 492 | 4240899 | ||
| BY209758.1 | - | 4 | 241 | |||
| L21671.1 | - | 4 | 396 | |||

Eps8
Genomic Map