siRNA Details
| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007963.2 | NP_031989.2 | 4 | 1094 | ||
| GenBank mRNA | AJ001482.1 | - | 2 | 250 | ||
| AK033517.1 | - | 4 | 528 | |||
| AK051098.1 | - | 4 | 951 | |||
| AK085548.1 | - | 4 | 1095 | |||
| BC076620.1 | - | 4 | 1046 | |||
| BC139762.1 | - | 2 | 212 | 40109903 | ||
| JQ665270.1 | - | 2 | 242 | |||

Mecom
Genomic Map