siRNA Details
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007984.2 | NP_032010.2 | 1 | 843 | ||
GenBank mRNA | AK083935.1 | - | 1 | 844 | ||
AK167670.1 | - | 1 | 843 | |||
BC010338.1 | - | 1 | 829 | |||
BC029281.1 | - | 1 | 831 | |||
BC037137.1 | - | 1 | 840 | |||
BC052408.1 | - | 1 | 832 | 5716179 | ||
L33726.1 | - | 1 | 827 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
