siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 2 | 432 | ||
| NM_007985.3 | NP_032011.2 | 2 | 432 | |||
| GenBank mRNA | AK035488.1 | - | 2 | 186 | ||
| AK042226.1 | - | 2 | 400 | |||
| AK080697.1 | - | 412 | ||||
| AK147025.1 | - | 2 | 412 | |||
| AK163161.1 | - | 2 | 433 | |||
| AK196042.1 | - | 2 | 162 | |||
| L08266.1 | - | 2 | 413 | |||

Fancc
Genomic Map