siRNA Details
| Gene Symbol: Fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037997.3 | NP_001033086.2 | 10 | 1387 | ||
| NM_001286415.1 | NP_001273344.1 | 9 | 1215 | |||
| NM_008000.2 | NP_032026.2 | 5 | 672 | |||
| GenBank mRNA | AF286537.1 | - | 9 | 1253 | ||
| AK082799.1 | - | 7 | 883 | |||
| AK157828.1 | - | 10 | 1354 | |||
| BC051249.1 | - | 2 | 131 | |||
| BC058100.1 | - | 9 | 1215 | 6813507 | ||
| M32054.1 | - | 5 | 672 | |||
| U76762.1 | - | 9 | 1320 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106928, XM_006245600, XM_008767372, XM_008767370, XM_008767369, and others.

Fer
Genomic Map