siRNA Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036627.1 | - | 9 | 3510 | ||
| AK041525.1 | - | 10 | 3657 | |||
| AK087638.1 | - | 9 | 3505 | |||
| DV662355.1 | - | 1 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esr1| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036627.1 | - | 9 | 3510 | ||
| AK041525.1 | - | 10 | 3657 | |||
| AK087638.1 | - | 9 | 3505 | |||
| DV662355.1 | - | 1 | 362 | |||