siRNA Details
| Gene Symbol: Esx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007957.2 | NP_031983.2 | 5 | 1435 | ||
| GenBank mRNA | AF004211.1 | - | 4 | 1523 | ||
| AF017734.1 | - | 4 | 1152 | |||
| AF017735.1 | - | 5 | 1235 | |||
| AF085715.1 | - | 5 | 1436 | |||

Esx1
Genomic Map