siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 2 | 473 | ||
| NM_007985.3 | NP_032011.2 | 2 | 473 | |||
| GenBank mRNA | AK035488.1 | - | 2 | 227 | ||
| AK042226.1 | - | 2 | 441 | |||
| AK080697.1 | - | 453 | ||||
| AK147025.1 | - | 2 | 453 | |||
| AK163161.1 | - | 2 | 474 | |||
| AK196042.1 | - | 2 | 203 | |||
| L08266.1 | - | 2 | 454 | |||

Fancc
Genomic Map