siRNA Details
Gene Symbol: Fgf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008003.2 | NP_032029.1 | 3 | 1798 | ||
GenBank mRNA | AF007268.1 | - | 3 | 1798 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008003.2 | NP_032029.1 | 3 | 1798 | ||
GenBank mRNA | AF007268.1 | - | 3 | 1798 |