siRNA Details
Gene Symbol: Akr1b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008012.1 | NP_032038.1 | 10 | 1268 | ||
GenBank mRNA | BC005789.1 | - | 10 | 1257 | 3603424 | |
U04204.1 | - | 10 | 1268 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Akr1b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008012.1 | NP_032038.1 | 10 | 1268 | ||
GenBank mRNA | BC005789.1 | - | 10 | 1257 | 3603424 | |
U04204.1 | - | 10 | 1268 |