siRNA Details
| Gene Symbol: Fpr-rs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008040.2 | NP_032066.2 | 1 | 436 | ||
| GenBank mRNA | BC134352.1 | - | 1 | 431 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpr-rs3| Gene Symbol: Fpr-rs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008040.2 | NP_032066.2 | 1 | 436 | ||
| GenBank mRNA | BC134352.1 | - | 1 | 431 | ||
Set Membership: |
|