siRNA Details
| Gene Symbol: Fxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113188.1 | NP_001106659.1 | 3 | 165 | ||
| NM_001113189.1 | NP_001106660.1 | 3 | 165 | |||
| NM_008053.2 | NP_032079.1 | 3 | 165 | |||
| GenBank mRNA | BC019139.1 | - | 3 | 146 | 5041635 | |
| CJ086949.1 | - | 3 | 166 | |||
| X90875.1 | - | 3 | 144 | |||

Fxr1
Genomic Map