siRNA Details
| Gene Symbol: Gabrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008069.4 | NP_032095.1 | 2 | 147 | ||
| GenBank mRNA | BC130258.1 | - | 2 | 151 | 40142758 | |
| BC157920.1 | - | 2 | 213 | 9007350 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012956, X15466.

Gabrb1
Genomic Map