siRNA Details
| Gene Symbol: Gas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008087.2 | NP_032113.1 | 3 | 479 | ||
| GenBank mRNA | AK014509.1 | - | 5 | 984 | ||
| AK142903.1 | - | 5 | 658 | |||
| BC013456.1 | - | 2 | 247 | 4237356 | ||
| BC053446.1 | - | 3 | 528 | 6336752 | ||
| M21828.1 | - | 3 | 479 | |||

Gas2
Genomic Map