siRNA Details
| Gene Symbol: Gata4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008092.3 | NP_032118.2 | 3 | 1252 | ||
| GenBank mRNA | AB075549.1 | - | 3 | 1270 | ||
| AF179424.1 | - | 2 | 673 | |||
| AK134639.1 | - | 2 | 276 | |||
| BC137824.1 | - | 2 | 1005 | 8860843 | ||
| M98339.1 | - | 3 | 1220 | |||
| U85046.1 | - | 3 | 1223 | |||

Gata4
Genomic Map