siRNA Details
| Gene Symbol: Gbas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008095.4 | NP_032121.3 | 2 | 222 | ||
| GenBank mRNA | AJ001261.1 | - | 2 | 200 | ||
| AK170336.1 | - | 2 | 168 | |||
| BC055893.1 | - | 2 | 171 | 4023066 | ||
| BY292750.1 | - | 2 | 223 | |||

Gbas
Genomic Map