siRNA Details
| Gene Symbol: Gcdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044744.1 | NP_001038209.2 | 4 | 371 | ||
| NM_008097.2 | NP_032123.3 | 3 | 428 | |||
| GenBank mRNA | AK165406.1 | - | 3 | 254 | ||
| AU080952.1 | - | 4 | 228 | |||
| BC061158.1 | - | 3 | 258 | 30314273 | ||
| BG807173.1 | - | 3 | 424 | |||
| BY301880.1 | - | 4 | 372 | |||
| U18992.1 | - | 3 | 209 | |||

Gcdh
Genomic Map