siRNA Details
| Gene Symbol: Flot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040403.1 | NP_001035493.1 | 7 | 728 | ||
| NM_001284227.1 | NP_001271156.1 | 8 | 812 | |||
| NM_001284228.1 | NP_001271157.1 | 8 | 675 | |||
| NM_008028.2 | NP_032054.1 | 8 | 819 | |||
| GenBank mRNA | AK004589.1 | - | 7 | 666 | ||
| AK030199.1 | - | 7 | 691 | |||
| AK149530.1 | - | 8 | 750 | |||
| AK170557.1 | - | 7 | 729 | |||
| AK208907.1 | - | 1 | 29 | |||
| BC070423.1 | - | 8 | 777 | 6403816 | ||
| U07890.1 | - | 8 | 689 | |||

Flot2
Genomic Map