siRNA Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008069.4 | NP_032095.1 | 4 | 287 | ||
GenBank mRNA | BC130258.1 | - | 4 | 291 | 40142758 | |
BC157920.1 | - | 4 | 353 | 9007350 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008069.4 | NP_032095.1 | 4 | 287 | ||
GenBank mRNA | BC130258.1 | - | 4 | 291 | 40142758 | |
BC157920.1 | - | 4 | 353 | 9007350 |