siRNA Details
| Gene Symbol: Gas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008087.2 | NP_032113.1 | 4, 5 | 657 | ||
| GenBank mRNA | AK014509.1 | - | 6, 7 | 1162 | ||
| AK142903.1 | - | 6, 7 | 836 | |||
| AK188527.1 | - | 1, 2 | 166 | |||
| BC013456.1 | - | 3, 4 | 425 | 4237356 | ||
| BC053446.1 | - | 4, 5 | 706 | 6336752 | ||
| M21828.1 | - | 4, 5 | 657 | |||

Gas2
Genomic Map