siRNA Details
| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008118.3 | NP_032144.2 | 5 | 708 | ||
| GenBank mRNA | AK078933.1 | - | 5 | 709 | ||
| BC118519.1 | - | 5 | 694 | 40105765 | ||
| L24191.1 | - | 5 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gif| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008118.3 | NP_032144.2 | 5 | 708 | ||
| GenBank mRNA | AK078933.1 | - | 5 | 709 | ||
| BC118519.1 | - | 5 | 694 | 40105765 | ||
| L24191.1 | - | 5 | 620 | |||