siRNA Details
| Gene Symbol: Gip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008119.2 | NP_032145.2 | 4 | 404 | ||
| GenBank mRNA | AK008308.1 | - | 4 | 404 | ||
| AK008525.1 | - | 3 | 260 | |||
| BC104314.1 | - | 4 | 394 | 40044479 | ||
| BC104315.1 | - | 4 | 394 | 40044480 | ||
| U34295.1 | - | 3 | 307 | |||

Gip
Genomic Map