siRNA Details
| Gene Symbol: Fpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008039.2 | NP_032065.1 | 2 | 709 | ||
| GenBank mRNA | AK054286.1 | - | 3 | 298 | ||
| BC117066.1 | - | 1 | 645 | 40126385 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpr2| Gene Symbol: Fpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008039.2 | NP_032065.1 | 2 | 709 | ||
| GenBank mRNA | AK054286.1 | - | 3 | 298 | ||
| BC117066.1 | - | 1 | 645 | 40126385 | ||
Set Membership: |
|