siRNA Details
| Gene Symbol: Igfbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159518.1 | NP_001152990.1 | 3, 4 | 717 | ||
| NM_008048.3 | NP_032074.3 | 2, 3 | 624 | |||
| GenBank mRNA | AB012886.1 | - | 2, 3 | 625 | ||
| AK002277.1 | - | 2, 3 | 620 | |||
| AK148749.1 | - | 2, 3 | 621 | |||
| AK156315.1 | - | 2, 3 | 626 | |||
| BC047202.1 | - | 2, 3 | 536 | |||
| BC092538.1 | - | 2, 3 | 620 | 30284911 | ||
| BF471478.1 | - | 3, 4 | 231 | |||
| L75822.2 | - | 2, 3 | 603 | |||

Igfbp7
Genomic Map