siRNA Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008066.3 | NP_032092.1 | 7, 8 | 1002 | ||
| GenBank mRNA | AK039055.1 | - | 7, 8 | 1003 | ||
| AK046953.1 | - | 2, 3 | 1136 | |||
| AK048165.1 | - | 900 | ||||
| BC115727.1 | - | 7, 8 | 924 | 40039253 | ||
| M86567.1 | - | 7, 8 | 753 | |||

Gabra2
Genomic Map