siRNA Details
| Gene Symbol: Gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038701.2 | NP_001033790.1 | 9 | 1777 | ||
| NM_008071.3 | NP_032097.1 | 9 | 1829 | |||
| GenBank mRNA | AK039070.1 | - | 9 | 1799 | ||
| AK083051.1 | - | 9 | 1842 | |||
| AK083107.1 | - | 9 | 1778 | |||
| AK140718.1 | - | 5 | 1373 | |||
| AK147247.1 | - | 9 | 1783 | |||

Gabrb3
Genomic Map