siRNA Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008078.2 | NP_032104.2 | 14 | 1812 | ||
| GenBank mRNA | AF326549.1 | - | 14 | 1476 | ||
| AF326550.1 | - | 14 | 1476 | |||
| BC018380.1 | - | 14 | 1527 | 4482097 | ||
| D42051.1 | - | 14 | 1828 | |||
| KC134211.1 | - | 14 | 1476 | |||
| L16980.1 | - | 14 | 1548 | |||

Gad2
Genomic Map