siRNA Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109657.3 | NP_001103127.1 | 14 | 2799 | ||
| NM_001277079.1 | NP_001264008.1 | 14 | 2628 | |||
| NM_001277080.1 | NP_001264009.1 | 13 | 2542 | |||
| NM_008088.5 | NP_032114.2 | 14 | 2605 | |||
| GenBank mRNA | AK076926.1 | - | 15 | 2683 | ||
| AK152149.1 | - | 12 | 2302 | |||
| AK154463.1 | - | 14 | 2584 | |||
| AK172943.1 | - | 10 | 2405 | |||
| U19860.1 | - | 14 | 2615 | |||

Gas7
Genomic Map